Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Polished, Glass Agitator, Four-Blade, 525mm OAL, for 1000mL Reactors, 67mm OD Agitator
SDP
Catalog No. CG2079B03
Encompass_Preferred
Stirring Shaft, 10mm, Polished, Glass Agitator, Four-Blade, 525mm OAL, for 1000mL Reactors, 67mm OD Agitator. Shaft is precision ground and polished to fit our CG-2076 and CG-2077 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The straight glass blades are spaced 90° apart and angled 15°. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.
Catalog No. CG2079B03 Supplier Chemglass Life Sciences Supplier No. CG2079B03
Stirring Shaft, 10mm, Polished, Glass Agitator, Four-Blade, 525mm OAL, for 1000mL Reactors, 67mm OD Agitator. Shaft is precision ground and polished to fit our CG-2076 and CG-2077 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The straight glass blades are spaced 90° apart and angled 15°. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.