Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Clockwise, Polished, Glass Agitator, Three Blade, 400mm OAL, Agitator OD 42mm, for use with 100-250mL Reactors
SDP
Catalog No. 501531435
Encompass_Preferred
Stirring Shaft, 10mm, Polished, Glass Agitator, Four-Blade, 460mm OAL, for 500mL Reactors, 67mm OD Agitator. Shaft is precision ground and polished to fit Chemglass's CG-2076 and CG-2077 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The straight glass blades are spaced 90 apart and angled 15. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.
Catalog No. 50-153-1435 Supplier Chemglass Life Sciences Supplier No. CG2079C50
Stirring Shaft, 10mm, Polished, Glass Agitator, Four-Blade, 460mm OAL, for 500mL Reactors, 67mm OD Agitator. Shaft is precision ground and polished to fit Chemglass's CG-2076 and CG-2077 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The straight glass blades are spaced 90 apart and angled 15. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.