Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Clockwise, Polished, Glass Agitator, Three Blade, 400mm OAL, Agitator OD 50mm, for use with 100-250mL Reactors
SDP
Catalog No. 501531436
Encompass_Preferred
Stirring Shaft, 10mm, Clockwise, Polished, Glass Agitator, Three Blade, 400mm OAL, Agitator OD 50mm, for use with 100-250mL Reactors. Shaft is precision ground and polished to fit Chemglass's CG-2077-H, CG-2077-G and CG-2077-U style 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120 apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR CLOCKWISE ROTATION.
Catalog No. 50-153-1436 Supplier Chemglass Life Sciences Supplier No. CG2079C52
Stirring Shaft, 10mm, Clockwise, Polished, Glass Agitator, Three Blade, 400mm OAL, Agitator OD 50mm, for use with 100-250mL Reactors. Shaft is precision ground and polished to fit Chemglass's CG-2077-H, CG-2077-G and CG-2077-U style 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120 apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR CLOCKWISE ROTATION.