Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Clockwise, Polished, Glass Agitator, Three Blade, 460mm OAL, Agitator OD 66mm, for use with 500mL Reactors
SDP
Catalog No. 501531438
Encompass_Preferred
Stirring Shaft, 10mm, Clockwise, Polished, Glass Agitator, Three Blade, 460mm OAL, Agitator OD 66mm, for use with 500mL Reactors. Shaft is precision ground and polished to fit Chemglass's CG-2077-H, CG-2077-G and CG-2077-U style 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120 apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR CLOCKWISE ROTATION.
Catalog No. 50-153-1438 Supplier Chemglass Life Sciences Supplier No. CG2079C56
Stirring Shaft, 10mm, Clockwise, Polished, Glass Agitator, Three Blade, 460mm OAL, Agitator OD 66mm, for use with 500mL Reactors. Shaft is precision ground and polished to fit Chemglass's CG-2077-H, CG-2077-G and CG-2077-U style 10mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120 apart and angled upward. PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR CLOCKWISE ROTATION.