Learn More
Ace Glass, Inc. Stir shaft with PTFE retreat agitator, 10mm OD x 400mm length PTFE covered stainless shaft with 50mm exposed for stir motor chuck, 50mm OD agitator with 10mm high blades
SDP
Catalog No. 50978130
Encompass
10mm O.D. x 400mm L, PTFE-covered stainless steel stir shaft with 50mm exposed for stir motor chuck. Shaft has a solid 50mm O.D. PTFE 30 retreat curve agitator with 10mm high blades.
Catalog No. 50-978-130 Supplier Ace Glass, Inc. Supplier No. 1385001
10mm O.D. x 400mm L, PTFE-covered stainless steel stir shaft with 50mm exposed for stir motor chuck. Shaft has a solid 50mm O.D. PTFE 30 retreat curve agitator with 10mm high blades.