Learn More
Ace Glass, Inc. Stir shaft with PTFE four blade screw propeller, 10mm ODx400mmlength PTFE covered stainless shaft with 50mm exposed for stir motor chuck, 50mm OD propeller with 22mm high blades
SDP
Catalog No. 50983549
Encompass
Stir shaft with PTFE four blade screw propeller, 10mm ODx400mmlength PTFE covered stainless shaft with 50mm exposed for stir motor chuck, 50mm OD propeller with 22mm high blades
Catalog No. 50-983-549 Supplier Ace Glass, Inc. Supplier No. 1385210
Stir shaft with PTFE four blade screw propeller, 10mm ODx400mmlength PTFE covered stainless shaft with 50mm exposed for stir motor chuck, 50mm OD propeller with 22mm high blades