Learn More
Metrohm USA STIR SHAFT
SDP
Supplier:  Metrohm USA 61909010
Encompass
Propeller stirrer, length from lower edge of SGJ: 96 mm. For use with beakers with 722, 802 Rod Stirrer
Catalog No. NC0415188