Learn More
Ace Glass, Inc. Stir shaft with PTFE four blade screw propeller, 19mm ODx900mmlength PTFE covered stainless shaft with 50mm exposed for stir motor chuck, 95mm OD propeller with 22mm high blades
SDP
Catalog No. 50978133
Encompass
Stir shaft with PTFE four blade screw propeller, 19mm ODx900mmlength PTFE covered stainless shaft with 50mm exposed for stir motor chuck, 95mm OD propeller with 22mm high blades
Catalog No. 50-978-133 Supplier Ace Glass, Inc. Supplier No. 1385219
Stir shaft with PTFE four blade screw propeller, 19mm ODx900mmlength PTFE covered stainless shaft with 50mm exposed for stir motor chuck, 95mm OD propeller with 22mm high blades