Learn More
Chemglass Life Sciences Stirring Shaft, 19mm, Polished, Glass Agitator, Three-Blade, 625mm OAL, for 3000mL Reactors, 105mm OD Agitator
SDP
Catalog No. CG2088A02
Encompass_Preferred
Stirring Shaft, 19mm, Polished, Glass Agitator, Three-Blade, 625mm OAL, for 3000mL Reactors, 105mm OD Agitator. Shaft is precision ground and polished to fit our CG-2086 19mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120° apart and angled upward.PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.
Catalog No. CG2088A02 Supplier Chemglass Life Sciences Supplier No. CG2088A02
Stirring Shaft, 19mm, Polished, Glass Agitator, Three-Blade, 625mm OAL, for 3000mL Reactors, 105mm OD Agitator. Shaft is precision ground and polished to fit our CG-2086 19mm PTFE bearings. Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120° apart and angled upward.PLEASE NOTE: STIRRER SHAFTS ARE DESIGNED FOR COUNTER-CLOCKWISE ROTATION.ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.