Learn More
Wilmad Labglass STIR SHAFT GLAS 10MM 12L
Supplier:  Wilmad Labglass LG9500102
Encompass_Preferred
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 515mm; Fits Flask Capacity: 12000mL. Precision ground with side button for attachment of Stirrer blades
Catalog No. 50-807-539