Learn More
Wilmad Labglass STIR SHAFT GLAS 10MM 22L
Supplier:  Wilmad Labglass LG9500104
Encompass_Preferred
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 590mm; Fits Flask Capacity: 22000mL. Precision ground with side button for attachment of Stirrer blades
Catalog No. 50-807-474