Learn More
Wilmad Labglass STIR SHAFT GLAS HD 22L
Catalog No. 50803934
Encompass_Preferred
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 19mm; Overall Length: 720mm; Fits Flask Capacity: 22000mL. Precision ground with side button for attaching LG-9515-series or LG-9516T-series Stirrer blades.
Catalog No. 50-803-934 Supplier Wilmad Labglass Supplier No. LG9504108
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 19mm; Overall Length: 720mm; Fits Flask Capacity: 22000mL. Precision ground with side button for attaching LG-9515-series or LG-9516T-series Stirrer blades.