Learn More
Wilmad Labglass STIR SHAFT GLAS POL 100-5000ML
Catalog No. 50803737
Encompass_Preferred
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 445mm; Flask Capacity: 100-5000mL. Precision ground and polished with side button for attachment of Stirrer blades.
Catalog No. 50-803-737 Supplier Wilmad Labglass Supplier No. LG9501100
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 445mm; Flask Capacity: 100-5000mL. Precision ground and polished with side button for attachment of Stirrer blades.