Learn More
Wilmad Labglass STIR SHAFT GLAS POL 12L
Catalog No. 50806848
Encompass_Preferred
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 515mm; Flask Capacity: 12000mL. Precision ground and polished with side button for attachment of Stirrer blades.
Catalog No. 50-806-848 Supplier Wilmad Labglass Supplier No. LG9501102
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 515mm; Flask Capacity: 12000mL. Precision ground and polished with side button for attachment of Stirrer blades.