Learn More
Wilmad Labglass STIR SHAFT GLAS POL 22L
Catalog No. 50806634
Encompass_Preferred
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 590mm; Flask Capacity: 22000mL. Precision ground and polished with side button for attachment of Stirrer blades.
Catalog No. 50-806-634 Supplier Wilmad Labglass Supplier No. LG9501104
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 590mm; Flask Capacity: 22000mL. Precision ground and polished with side button for attachment of Stirrer blades.