Learn More
Wilmad Labglass STIR SHAFT GLS 10MM 100-5000ML
Catalog No. 50803271
Encompass_Preferred
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 310mm; Fits Flask Capacity: 100-5000mL. Precision ground with side button for attachment of Stirrer blades
Catalog No. 50-803-271 Supplier Wilmad Labglass Supplier No. LG9500100
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 10mm; Overall Length: 310mm; Fits Flask Capacity: 100-5000mL. Precision ground with side button for attachment of Stirrer blades