Learn More
Chemglass Life Sciences Stirring Shaft, PTFE Covered SS, 525mm OAL, 66mm OD Retreating Blade, Fits Reactors up to 1000mL
SDP
Catalog No. NC0814333
Encompass_Preferred
Stirring Shaft, PTFE Covered SS, 525mm OAL, 66mm OD Retreating Blade, Fits Reactors up to 1000mL
Catalog No. NC0814333 Supplier Chemglass Life Sciences Supplier No. CG2079X100
Stirring Shaft, PTFE Covered SS, 525mm OAL, 66mm OD Retreating Blade, Fits Reactors up to 1000mL