Learn More
Chemglass Life Sciences Stirring Shaft, PTFE Covered SS, 625mm OAL, 4-Blade, 127mm OD Blades, High Viscosity Style, Fits 3-5L Reactors
SDP
Catalog No. 501311206
Encompass_Preferred
Stirring Shaft, PTFE Covered SS, 625mm OAL, 4-Blade, 127mm OD Blades, High Viscosity Style, Fits 3-5L Reactors. 16mm O.D. shaft is constructed using a molding process whereas the blade is molded directly onto the PTFE coated stainless steel shaft. This makes the stirrer chemically inert, easy to clean and can be used up to 280°C. The top of the shaft has 50mm of exposed 1/2" diameter stainless steel for rigid clamping to a coupling such as CG-2044-02, CG-2044-05 or CG-2046-03.Note: Shaft must be used with CG-2077-U or CG-2077-M style bearing
Catalog No. 50-131-1206 Supplier Chemglass Life Sciences Supplier No. CG2096X500
Stirring Shaft, PTFE Covered SS, 625mm OAL, 4-Blade, 127mm OD Blades, High Viscosity Style, Fits 3-5L Reactors. 16mm O.D. shaft is constructed using a molding process whereas the blade is molded directly onto the PTFE coated stainless steel shaft. This makes the stirrer chemically inert, easy to clean and can be used up to 280°C. The top of the shaft has 50mm of exposed 1/2" diameter stainless steel for rigid clamping to a coupling such as CG-2044-02, CG-2044-05 or CG-2046-03.Note: Shaft must be used with CG-2077-U or CG-2077-M style bearing