Learn More
Ace Glass, Inc. 10mm 42cm stainless steel stir shaft, polished, removable button type, 10cm flat section
SDP
Catalog No. 509558205
Encompass
10mm O.D. 42cm stainless steel polished stir shaft, drilled type, with removable button. Stir Shaft also has a 10cm flat section for use with agitators secured by set screws. For use with Trubore and PTFE bearings.
Catalog No. 50-955-8205 Supplier Ace Glass, Inc. Supplier No. 807402
10mm O.D. 42cm stainless steel polished stir shaft, drilled type, with removable button. Stir Shaft also has a 10cm flat section for use with agitators secured by set screws. For use with Trubore and PTFE bearings.