Learn More
Wilmad Labglass STIR SHAFT SSTEEL 100-5000ML
Catalog No. 50800457
Encompass
Stirrer Shaft, Button Style, Polished, Stainless Steel. Shaft O.D.: 10mm, Overall Length: 515mm; Fits Flask Capacity: 100-5000mL. Precision ground and polished with side button for attachment of Stirrer blades.
Catalog No. 50-800-457 Supplier Wilmad Labglass Supplier No. LG9502100
Stirrer Shaft, Button Style, Polished, Stainless Steel. Shaft O.D.: 10mm, Overall Length: 515mm; Fits Flask Capacity: 100-5000mL. Precision ground and polished with side button for attachment of Stirrer blades.