Learn More
Ace Glass, Inc. 10mm 44cm solid glass stir shaft, PTFE covered, button type
SDP
Catalog No. 509558201
Encompass_Preferred
44cm, 10mm O.D. button type, precision ground glass stirring shafts with PTFE coating that permits higher stirring speeds with Trubore bearings.
Catalog No. 50-955-8201 Supplier Ace Glass, Inc. Supplier No. 807005
44cm, 10mm O.D. button type, precision ground glass stirring shafts with PTFE coating that permits higher stirring speeds with Trubore bearings.