Learn More
V&P Scientific Inc STIR ELEMENT, SANDWICH STYLE TUNING FORK - WITH PIVOT POINT WINGS, NdFeB Magnet, 48MGO, PTFE Plate, Used in Tubes/Vials, 10/pack
Catalog No. NC2212071
Encompass
STIR ELEMENT, SANDWICH STYLE TUNING FORK - WITH PIVOT POINT WINGS, NdFeB Magnet, 48MGO, PTFE Plate, 35.0mm Length, 10.0mm Width, 1.6mm Height, 80°C Max Operating Temp for Magnets
This item is not returnable. View return policy
Catalog No. NC2212071 Supplier V&P Scientific Inc Supplier No. VP 772FN1035TF
STIR ELEMENT, SANDWICH STYLE TUNING FORK - WITH PIVOT POINT WINGS, NdFeB Magnet, 48MGO, PTFE Plate, 35.0mm Length, 10.0mm Width, 1.6mm Height, 80°C Max Operating Temp for Magnets