Learn More
V&P Scientific Inc STIR ELEMENT, RECTANGLE, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, Used in Bottles, 10/pack
Catalog No. NC0111276
Encompass
STIR ELEMENT, RECTANGLE, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, 25.0mm Length, 5.5mm Width, 5.5mm Height, 260°C Max Operating Temp for Coating
Catalog No. NC0111276 Supplier V&P Scientific Inc Supplier No. VP 77825
STIR ELEMENT, RECTANGLE, AlNiCo Magnet, 5.5MGO, PTFE Encapsulated, 25.0mm Length, 5.5mm Width, 5.5mm Height, 260°C Max Operating Temp for Coating