Learn More
Andwin Scientific Stir bars egg-shaped, Spinbar, 1 each
SDP
Catalog No. NC9011181
Encompass_Preferred
Spinbar, Stir bars egg-shaped, Designed specifically for round bottom Flasks and vessels. 19L x 9.5mm dia., For round-bottom Flasks sized 10, 25, 50 and 100mL, 1/EA
Catalog No. NC9011181 Supplier Andwin Scientific Supplier No. 58949006
Spinbar, Stir bars egg-shaped, Designed specifically for round bottom Flasks and vessels. 19L x 9.5mm dia., For round-bottom Flasks sized 10, 25, 50 and 100mL, 1/EA