Learn More
Ace Glass, Inc. Stirbar, PTFE, egg shape, 25.4mmx12.7mm
SDP
Catalog No. 50983532
Encompass
25.4mm x 12.7mm egg-shaped PTFE magnetic stir bar which provides a pivot point without a separate ring or abrupt change in contour. Designed to be used with round bottom flasks and DynaBlocs.
Catalog No. 50-983-532 Supplier Ace Glass, Inc. Supplier No. 1366305
25.4mm x 12.7mm egg-shaped PTFE magnetic stir bar which provides a pivot point without a separate ring or abrupt change in contour. Designed to be used with round bottom flasks and DynaBlocs.