Learn More
Andwin Scientific STIRBARS MICRO 1.5MMX8MM 12/CS
SDP
Catalog No. NC9956727
Encompass_Preferred
The Micro PTFE Stir Bars measure 8mm x 1.5mm (5/16 in. x 1/16 in.). This product is sold in a case of 12.
Catalog No. NC9956727 Supplier Andwin Scientific Supplier No. MX3288D
The Micro PTFE Stir Bars measure 8mm x 1.5mm (5/16 in. x 1/16 in.). This product is sold in a case of 12.