Learn More
V&P Scientific Inc STIR ELEMENT, DISC, NdFeB Magnet, 42MGO, PVDF Encapsulated, Used in Microplates, Syringes, Tubes/Vials, 10/pack
Catalog No. NC0493857
Encompass
STIR ELEMENT, DISC, NdFeB Magnet, 42MGO, PVDF Encapsulated, 8.0mm Diameter, 1.8mm Height, 150°C Max Operating Temp for Magnets
Catalog No. NC0493857 Supplier V&P Scientific Inc Supplier No. VP 772DPN4282
STIR ELEMENT, DISC, NdFeB Magnet, 42MGO, PVDF Encapsulated, 8.0mm Diameter, 1.8mm Height, 150°C Max Operating Temp for Magnets