Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1578021
Encompass_Preferred
Aluminum housed stirmantle for 500ml spherical flask, stir 50-750rpm, heat to 400 degrees C internal operating temperature. 270W 115V
Catalog No. NC1578021 Supplier Glas Col LLC Supplier No. 100D TEM106
Aluminum housed stirmantle for 500ml spherical flask, stir 50-750rpm, heat to 400 degrees C internal operating temperature. 270W 115V