Learn More
Cayman Chemical StIrIpentol 10mg
SDP
Catalog No. 502979800
Encompass_Preferred
An antiepileptic compound; positive allosteric modulator of GABAA receptors that potentiates GABA-mediated activation of α3 subunit-containing GABAA receptors; inhibits GABA reuptake in vitro and increases the release of GABA in neonatal rat hippocampal slices; inhibits LDH1 and LDH4 at 500 µM
Catalog No. 50-297-9800 Supplier Cayman Chemical Supplier No. 1778110 MG
An antiepileptic compound; positive allosteric modulator of GABAA receptors that potentiates GABA-mediated activation of α3 subunit-containing GABAA receptors; inhibits GABA reuptake in vitro and increases the release of GABA in neonatal rat hippocampal slices; inhibits LDH1 and LDH4 at 500 µM