Learn More
TARGETMOL CHEMICALS INC STIRIPENTOL 25MG
SDP
Catalog No. 502579875
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Stiripentol (BCX2600) (STP) is an anticonvulsant agent which can inhibit N-demethylation of CLB to NCLB mediated by CYP3A4 (noncompetitively) and CYP2C19 (competitively) with Ki of 1.59/0.516 uM and IC50 of 1.58/3.29 uM respectively. purity: 99%
Catalog No. 50-257-9875 Supplier TARGETMOL CHEMICALS INC Supplier No. T449825MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Stiripentol (BCX2600) (STP) is an anticonvulsant agent which can inhibit N-demethylation of CLB to NCLB mediated by CYP3A4 (noncompetitively) and CYP2C19 (competitively) with Ki of 1.59/0.516 uM and IC50 of 1.58/3.29 uM respectively. purity: 99%