Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stiripentol | 49763-96-4 | MFCD00869310 | >=98.0% | 234.29 g/mol | C14H18O3 | 200 MG
SDP

Catalog No. 5000244566
Encompass_Preferred

Stiripentol is an anticonvulsant research compound used as adjunct therapy in seizure disorders such as Dravet syndrome. This product listing represents a 200 mg research reagent with analytical identifiers and purity information available from supplier and reference databases.

  • High purity ≥98.0% (HPLC).
  • Molecular formula C14H18O3.
  • Molecular weight 234.29 g/mol.
  • Pack contains 200 mg of compound.
  • For research use only.

Catalog No. 50-002-44566 Supplier Medchemexpress LLC Supplier No. HY103392200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stiripentol is an anticonvulsant research compound used as adjunct therapy in seizure disorders such as Dravet syndrome. This product listing represents a 200 mg research reagent with analytical identifiers and purity information available from supplier and reference databases.

  • High purity ≥98.0% (HPLC).
  • Molecular formula C14H18O3.
  • Molecular weight 234.29 g/mol.
  • Pack contains 200 mg of compound.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.