Learn More
Cayman Chemical Stiripentol, Purity: ≥98%, CAS: 49763-96-4, 50 mg
SDP
Catalog No. 502980419
Encompass_Preferred
An antiepileptic compound; positive allosteric modulator of GABAA receptors that potentiates GABA-mediated activation of α3 subunit-containing GABAA receptors; inhibits GABA reuptake in vitro and increases the release of GABA in neonatal rat hippocampal slices; inhibits LDH1 and LDH4 at 500 µM
Catalog No. 50-298-0419 Supplier Cayman Chemical Supplier No. 1778150 MG
An antiepileptic compound; positive allosteric modulator of GABAA receptors that potentiates GABA-mediated activation of α3 subunit-containing GABAA receptors; inhibits GABA reuptake in vitro and increases the release of GABA in neonatal rat hippocampal slices; inhibits LDH1 and LDH4 at 500 µM