Learn More
Azzota Corp Stirring Heating Mantle, Capacity: 100Ml , Maximum Temp. Is 716'F (380'C). Stir Speed: 0 To 1400Rpm
Catalog No. 501179601
Encompass_Preferred
Stirring Heating Mantle, Capacity: 100ml , Maximum temp. is 716'F (380'C). Stir speed: 0 to 1400rpm
Catalog No. 50-117-9601 Supplier Azzota Corp Supplier No. SHM100
Stirring Heating Mantle, Capacity: 100ml , Maximum temp. is 716'F (380'C). Stir speed: 0 to 1400rpm