Learn More
Chemglass Life Sciences Plain PTFE Magnetic Stirring Bar, 50 x 8 mm, for Magnetic Stirrers
SDP
Catalog No. NC3304525
Encompass_Preferred
Plain PTFE-coated magnetic stirring bar (50 x 8 mm) for general-purpose laboratory mixing on magnetic stir plates. Its cylindrical plain profile provides effective agitation and produces increased turbulence at low speeds, while the PTFE coating offers chemical resistance and a non-stick surface.
  • Plain cylindrical shape for general-purpose stirring.
  • Dimensions: 50 x 8 mm.
  • PTFE coating provides chemical resistance and non-stick surface.
  • Produces increased turbulence at low stirrer speeds.
  • Compatible with standard magnetic stir plates and laboratory mixers.
  • Sold individually for easy replacement.
Catalog No. NC3304525 Supplier Chemglass Life Sciences Supplier No. CG2001P07
Plain PTFE-coated magnetic stirring bar (50 x 8 mm) for general-purpose laboratory mixing on magnetic stir plates. Its cylindrical plain profile provides effective agitation and produces increased turbulence at low speeds, while the PTFE coating offers chemical resistance and a non-stick surface.
  • Plain cylindrical shape for general-purpose stirring.
  • Dimensions: 50 x 8 mm.
  • PTFE coating provides chemical resistance and non-stick surface.
  • Produces increased turbulence at low stirrer speeds.
  • Compatible with standard magnetic stir plates and laboratory mixers.
  • Sold individually for easy replacement.