Learn More
Ace Glass, Inc. Stirrer, 10mm, 24/40, 250-500mL, complete with glass blade
SDP
Catalog No. 50975918
Encompass_Preferred
250-500ml Precision Trubore Stirrer Assembly. Consists of 24/40, 10mm I.D. bearing, bushing, precision ground, button type glass shaft and glass blade.
Catalog No. 50-975-918 Supplier Ace Glass, Inc. Supplier No. 804111
250-500ml Precision Trubore Stirrer Assembly. Consists of 24/40, 10mm I.D. bearing, bushing, precision ground, button type glass shaft and glass blade.