Learn More
Ace Glass, Inc. Stirrer, 10mm, 29/42, 500-1000mL, complete with glass blade
SDP
Catalog No. 50975919
Encompass_Preferred
500-1000ml Precision Trubore Stirrer Assembly. Consists of 29/42, 10mm I.D. bearing, bushing, precision ground, button type glass shaft and glass blade.
Catalog No. 50-975-919 Supplier Ace Glass, Inc. Supplier No. 804115
500-1000ml Precision Trubore Stirrer Assembly. Consists of 29/42, 10mm I.D. bearing, bushing, precision ground, button type glass shaft and glass blade.