Learn More
Ace Glass, Inc. Stirrer, 10mm, 45/50, 5000-12000mL, complete with glass blade
SDP
Catalog No. 50979628
Encompass_Preferred
5000-12000ml Precision Trubore Stirrer Assembly. Consists of 45/50, 10mm I.D. bearing, bushing, precision ground, button type glass shaft and glass blade.
Catalog No. 50-979-628 Supplier Ace Glass, Inc. Supplier No. 804120
5000-12000ml Precision Trubore Stirrer Assembly. Consists of 45/50, 10mm I.D. bearing, bushing, precision ground, button type glass shaft and glass blade.