Learn More
Chemglass Life Sciences Stirrer, Arrow, Air Driven, General Purpose, 50-1200 RPM, 18.0 in-lbs Constant Torque, 7:1 Gear Ratio
SDP
Catalog No. CG202502
Encompass_Preferred
Stirrer, Arrow, Air Driven, General Purpose, 50-1200 RPM, 18.0 in-lbs Constant Torque, 7:1 Gear Ratio. Air driven stirrer for use when an electrical stirrer may be hazardous or impractical. Ideal for mixing all types of solvents, lacquers & volatile chemicals without the possibility of spark ignition. Motor accepts air input line pressure from 35 to 1000 PSI & delivers up to 1/3 Hp @ 80 PSI. A needle valve controls the mixing speed. A muffler ensures quiet operation. Supplied complete with a 9/16" Diameter x 10" long aluminum support rod, 4ft length of air hose w/ double fittings for connection to air supply, & a 3/8” diameter x 12” long stirrer shaft having a 2-1/2" diameter S.S. propellerCONSTANT TORQUE VALUES GIVEN AT A PRESSURE OF 80 PSI.PLEASE NOTE: FOR LONG LIFE OF YOUR AIR POWERED STIRRER, IT IS RECOMMENDED THAT THE AIR SUPPLY BE FILTERED TO REMOVE OIL AND WATER AEROSOLS. IN ADDITION, AN AUTOMATIC AIR LINE LUBRICATOR MUST BE INSTALLED IN THE AIR LINE JUST AHEAD OF THE MOTOR.
Catalog No. CG202502 Supplier Chemglass Life Sciences Supplier No. CG202502
Stirrer, Arrow, Air Driven, General Purpose, 50-1200 RPM, 18.0 in-lbs Constant Torque, 7:1 Gear Ratio. Air driven stirrer for use when an electrical stirrer may be hazardous or impractical. Ideal for mixing all types of solvents, lacquers & volatile chemicals without the possibility of spark ignition. Motor accepts air input line pressure from 35 to 1000 PSI & delivers up to 1/3 Hp @ 80 PSI. A needle valve controls the mixing speed. A muffler ensures quiet operation. Supplied complete with a 9/16" Diameter x 10" long aluminum support rod, 4ft length of air hose w/ double fittings for connection to air supply, & a 3/8” diameter x 12” long stirrer shaft having a 2-1/2" diameter S.S. propellerCONSTANT TORQUE VALUES GIVEN AT A PRESSURE OF 80 PSI.PLEASE NOTE: FOR LONG LIFE OF YOUR AIR POWERED STIRRER, IT IS RECOMMENDED THAT THE AIR SUPPLY BE FILTERED TO REMOVE OIL AND WATER AEROSOLS. IN ADDITION, AN AUTOMATIC AIR LINE LUBRICATOR MUST BE INSTALLED IN THE AIR LINE JUST AHEAD OF THE MOTOR.