Learn More
Chemglass Life Sciences Stirrer, Arrow, Light to Medium Duty, 0-6000 MAX RPM, 16.8 in-oz Constant Torque, 1/10 Hp Motor
SDP
Catalog No. CG202101
Encompass_Preferred
Stirrer, Arrow, Light to Medium Duty, 0-6000 MAX RPM, 16.8 in-oz Constant Torque, 1/10 Hp Motor. Variable high speed stirrer is ideal for light to medium-light viscosity fluids. The totally enclosed non-ventilated motor, keeps corrosive fumes and dust out for a long and trouble free service life. The permanent magnet DC motor provides maximum constant torque through entire speed range. The eye level heavy cast aluminum control box is integrated with the motor and has an on/off switch along with a speed control knob. Stirrer is protected from overload with a manually resettable circuit breaker. Electric stirrer is supplied complete with a 9/16" Diameter x 10" long aluminum support rod, 6ft 3-wire grounded plug, and a 3/8” diameter x 12” long stirrer shaft having a 2-1/2" diameter stainless steel propeller.
Catalog No. CG202101 Supplier Chemglass Life Sciences Supplier No. CG202101
Stirrer, Arrow, Light to Medium Duty, 0-6000 MAX RPM, 16.8 in-oz Constant Torque, 1/10 Hp Motor. Variable high speed stirrer is ideal for light to medium-light viscosity fluids. The totally enclosed non-ventilated motor, keeps corrosive fumes and dust out for a long and trouble free service life. The permanent magnet DC motor provides maximum constant torque through entire speed range. The eye level heavy cast aluminum control box is integrated with the motor and has an on/off switch along with a speed control knob. Stirrer is protected from overload with a manually resettable circuit breaker. Electric stirrer is supplied complete with a 9/16" Diameter x 10" long aluminum support rod, 6ft 3-wire grounded plug, and a 3/8” diameter x 12” long stirrer shaft having a 2-1/2" diameter stainless steel propeller.