Learn More
Chemglass Life Sciences 6mm Micro Stirrer Assembly, Complete with Shaft, Bearing and Blade, 14/20
SDP
Catalog No. CG206001
Encompass_Preferred
6mm Micro Stirrer Assembly, Complete with Shaft, Bearing and Blade, 14/20. Micro stirrer has a 6mm OD x 305mm long precision ground button style shaft and a football shaped 11 x 33mm TFE blade. The 11 x 33mm blade can be used in flasks having a capacity of 25mL and above. Complete stirrer assembly consists of a shaft, bearing and blade.
Catalog No. CG206001 Supplier Chemglass Life Sciences Supplier No. CG206001
6mm Micro Stirrer Assembly, Complete with Shaft, Bearing and Blade, 14/20. Micro stirrer has a 6mm OD x 305mm long precision ground button style shaft and a football shaped 11 x 33mm TFE blade. The 11 x 33mm blade can be used in flasks having a capacity of 25mL and above. Complete stirrer assembly consists of a shaft, bearing and blade.