Learn More
Chemglass Life Sciences 10mm Semi-Micro Stirrer Assembly, Complete with Shaft, Bearing and Blade, 19/22
SDP
Catalog No. CG206401
Encompass_Preferred
10mm Semi-Micro Stirrer Assembly, Complete with Shaft, Bearing and Blade, 19/22. Semi-micro stirrer has a 10mm OD x 440mm long precision ground button style shaft and a 15 x 46mm TFE blade. Lower end of shaft has a reduced diameter to allow passage through a 19/22 outer joint. The 15 x 46mm blade can be used in flasks having a capacity of 100mL and above. Complete stirrer assembly consists of a shaft, bearing and blade.
Catalog No. CG206401 Supplier Chemglass Life Sciences Supplier No. CG206401
10mm Semi-Micro Stirrer Assembly, Complete with Shaft, Bearing and Blade, 19/22. Semi-micro stirrer has a 10mm OD x 440mm long precision ground button style shaft and a 15 x 46mm TFE blade. Lower end of shaft has a reduced diameter to allow passage through a 19/22 outer joint. The 15 x 46mm blade can be used in flasks having a capacity of 100mL and above. Complete stirrer assembly consists of a shaft, bearing and blade.