Learn More
Chemglass Life Sciences Stir Bar, Magnetic, PTFE, Turbo, Rare Earth, Elliptical, 25 x 14mm
SDP
Catalog No. NC1781567
Encompass_Preferred
Exceptional strength coupled with almost total resistance to demagnetisation. Identified by an inert carbon black spot.
Catalog No. NC1781567 Supplier Chemglass Life Sciences Supplier No. CG2005T03
Exceptional strength coupled with almost total resistance to demagnetisation. Identified by an inert carbon black spot.