Learn More
Cole-Parmer Cole-Parmer Battery-Powered Magnetic Stirrer 04804-01
Supplier:  Cole-Parmer 0480401
Encompass_Preferred
Cole-Parmer Battery-Powered Magnetic Stirrer,Stepless speed control to 700 rpm,Take this compact, battery-powered magnetic stirrer anywhere you need vigorous samplestirring. Suitable for use inside incubators, fume hoods, or glove boxes. Or, use it in the lab and eliminate the need for cumbersome extension cords. Stirrer features stepless speed control knob, corrosion-resistant epoxy-coated metal case, nonskid rubber top plate, and four rubber feet.
Catalog No. NC9900446