Learn More
Chemglass Life Sciences Stirrer Bearing, PTFE, Chem-Stir, Complete, 19mm, 45/50 Inner Joint
SDP
Catalog No. CG208601
Encompass_Preferred
Stirrer Bearing, PTFE, Chem-Stir, Complete, 19mm, 45/50 Inner Joint. Grease-Free PTFE stirrer bearing designed for use under moderate vacuum applications. The bearing consists of a threaded glass adapter having an inner joint, a set of PTFE washer style bearings, a stainless steel spring, nylon alignment ring and Celcon compression cap. The unique design of this bearing provides hours of continuous vacuum operation, with a minimum of "flaking" of the PTFE bearing surfaces.PLEASE NOTE: FOR USE ONLY WITH CG-2087 and CG-2088 PRECISION GROUND AND POLISHED STIRRER SHAFTS.
Catalog No. CG208601 Supplier Chemglass Life Sciences Supplier No. CG208601
Stirrer Bearing, PTFE, Chem-Stir, Complete, 19mm, 45/50 Inner Joint. Grease-Free PTFE stirrer bearing designed for use under moderate vacuum applications. The bearing consists of a threaded glass adapter having an inner joint, a set of PTFE washer style bearings, a stainless steel spring, nylon alignment ring and Celcon compression cap. The unique design of this bearing provides hours of continuous vacuum operation, with a minimum of "flaking" of the PTFE bearing surfaces.PLEASE NOTE: FOR USE ONLY WITH CG-2087 and CG-2088 PRECISION GROUND AND POLISHED STIRRER SHAFTS.