Learn More
Chemglass Life Sciences Stirrer Bearing with side Hose Connection, PTFE, Complete, fits 10mm Shaft, 29/42 Inner Joint
SDP
Catalog No. CG2077A02
Encompass_Preferred
Stirrer Bearing with side Hose Connection, PTFE, Complete, fits 10mm Shaft, 29/42 Inner Joint. PTFE stirrer bearing with a hose connection for vacuum or inert gas source on a single neck flask. Grease-free design has only glass and PTFE in contact with the reaction, and can be used under moderate vacuum applications. Bearing consists of a threaded glass adapter having the listed inner joint size, a saddle o-ring, a body bushing with a compression and locking nut, and a precision PTFE inner bearing. Saddle o-ring provides vacuum tight seal and is replaceable at a fraction of the cost of the entire bearing. Hose connection is 10mm at the largest serration. PLEASE NOTE: FOR USE ONLY WITH CG-2078 OR CG-2079 PRECISION GROUND AND POLISHED STIRRER SHAFTS.
Catalog No. CG2077A02 Supplier Chemglass Life Sciences Supplier No. CG2077A02
Stirrer Bearing with side Hose Connection, PTFE, Complete, fits 10mm Shaft, 29/42 Inner Joint. PTFE stirrer bearing with a hose connection for vacuum or inert gas source on a single neck flask. Grease-free design has only glass and PTFE in contact with the reaction, and can be used under moderate vacuum applications. Bearing consists of a threaded glass adapter having the listed inner joint size, a saddle o-ring, a body bushing with a compression and locking nut, and a precision PTFE inner bearing. Saddle o-ring provides vacuum tight seal and is replaceable at a fraction of the cost of the entire bearing. Hose connection is 10mm at the largest serration. PLEASE NOTE: FOR USE ONLY WITH CG-2078 OR CG-2079 PRECISION GROUND AND POLISHED STIRRER SHAFTS.