Learn More
Chemglass Life Sciences Stirrer Bearing, Heavy Duty, for 19mm Shaft, with Tooled Lubricant Cup, 45/50 Inner Joint
SDP
Catalog No. 50873400
Encompass_Preferred
Stirrer Bearing, Heavy Duty, for 19mm Shaft, with Tooled Lubricant Cup, 45/50 Inner Joint. Heavy duty 19mm stirrer bearing for use in stirring large volumes under high torque. Bearing has a tooled lubricant cup. PLEASE NOTE: NOT FOR USE WITH CG-2087 or CG-2088 PRECISION GROUND AND POLISHED STIRRER SHAFTS.
Catalog No. 50-873-400 Supplier Chemglass Life Sciences Supplier No. CG208201
Stirrer Bearing, Heavy Duty, for 19mm Shaft, with Tooled Lubricant Cup, 45/50 Inner Joint. Heavy duty 19mm stirrer bearing for use in stirring large volumes under high torque. Bearing has a tooled lubricant cup. PLEASE NOTE: NOT FOR USE WITH CG-2087 or CG-2088 PRECISION GROUND AND POLISHED STIRRER SHAFTS.