Learn More
Chemglass Life Sciences Stirrer Bearing, Macro, Glass, with tooled Lubricant Cup, for 10mm Shaft, 45/50 Inner Joint
SDP
Catalog No. CG207105
Encompass_Preferred
Stirrer Bearing, Macro, Glass, with tooled Lubricant Cup, for 10mm Shaft, 45/50 Inner Joint. Similar to CG-2070 but with a tooled lubricant cup at the top of the bearing to hold an increased amount of lubricant. NOTE: Not for use with CG-2078 or CG-2079 polished stirrer shafts.
Catalog No. CG207105 Supplier Chemglass Life Sciences Supplier No. CG207105
Stirrer Bearing, Macro, Glass, with tooled Lubricant Cup, for 10mm Shaft, 45/50 Inner Joint. Similar to CG-2070 but with a tooled lubricant cup at the top of the bearing to hold an increased amount of lubricant. NOTE: Not for use with CG-2078 or CG-2079 polished stirrer shafts.