Learn More
Chemglass Life Sciences Stirrer Bearing, Macro, Glass, for 10mm Shaft, 29/42 Inner Joint
SDP
Catalog No. 50873394
Encompass_Preferred
Stirrer Bearing, Macro, Glass, for 10mm Shaft, 29/42 Inner Joint. Precision bore bearing for use in moderate vacuum with a CG-2074 or CG-2075 ground 10mm stirrer shaft. NOTE: Not for use with CG-2078 or CG-2079 polished stirrer shafts.
Catalog No. 50-873-394 Supplier Chemglass Life Sciences Supplier No. CG207002
Stirrer Bearing, Macro, Glass, for 10mm Shaft, 29/42 Inner Joint. Precision bore bearing for use in moderate vacuum with a CG-2074 or CG-2075 ground 10mm stirrer shaft. NOTE: Not for use with CG-2078 or CG-2079 polished stirrer shafts.