Learn More
Chemglass Life Sciences Stirrer Bearing, Heavy Duty, for 19mm Shaft, Jacketed, 45/50 Inner Joint
SDP
Catalog No. CG208301
Encompass_Preferred
Stirrer Bearing, Heavy Duty, for 19mm Shaft, Jacketed, 45/50 Inner Joint. Similar to CG-2082 but with a water jacket to control bearing temperature. For use in moderate vacuum with a CG-2084 or CG-2085 precision ground 19mm OD stirrer shaft. PLEASE NOTE: NOT FOR USE WITH CG-2087 or CG-2088 PRECISION GROUND AND POLISHED STIRRER SHAFTS.
Catalog No. CG208301 Supplier Chemglass Life Sciences Supplier No. CG208301
Stirrer Bearing, Heavy Duty, for 19mm Shaft, Jacketed, 45/50 Inner Joint. Similar to CG-2082 but with a water jacket to control bearing temperature. For use in moderate vacuum with a CG-2084 or CG-2085 precision ground 19mm OD stirrer shaft. PLEASE NOTE: NOT FOR USE WITH CG-2087 or CG-2088 PRECISION GROUND AND POLISHED STIRRER SHAFTS.