Learn More
Chemglass Life Sciences Stirrer Bearing, Macro, Jacketed, Glass, for 10mm Shaft, 45/50 Inner Joint
SDP
Catalog No. CG207204
Encompass_Preferred
Stirrer Bearing, Macro, Jacketed, Glass, for 10mm Shaft, 45/50 Inner Joint. Similar to CG-2070, but water jacketed to control bearing temperature. For use in moderate vacuum with a CG-2074 or CG-2075 ground 10mm stirrer shaft. NOTE: Not for use with CG-2078 or CG-2079 polished stirrer shafts.
Catalog No. CG207204 Supplier Chemglass Life Sciences Supplier No. CG207204
Stirrer Bearing, Macro, Jacketed, Glass, for 10mm Shaft, 45/50 Inner Joint. Similar to CG-2070, but water jacketed to control bearing temperature. For use in moderate vacuum with a CG-2074 or CG-2075 ground 10mm stirrer shaft. NOTE: Not for use with CG-2078 or CG-2079 polished stirrer shafts.